Human recombinant MMP2 (catalytic domain)
Fournisseur: ENZO LIFE SCIENCES
BMLSE2370010
ENZOBMLSE2370010EA 511.5 EUR InStock
ENZOBMLSE2370010
Human recombinant MMP2 (catalytic domain)
Protéines et Peptides
Produced in yeast. Active recombinant matrix metalloproteinase-2 (MMP-2, gelatinase A, 72kDa type IV collagenase) cloned from human cDNA.
The enzyme consists of residues Tyr110-Asp452 (NM_004530), which comprises the catalytic/fibronectin domain of human MMP-2, with a C-terminal purification tag. This represents a naturally occurring active form of MMP-2 which lacks the C-terminal hemopexin domain.
MMPs lacking this domain cannot cleave native collagens; however, activity toward other targets such as gelatin, casein, or peptide substrates is unaffected.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité