Human recombinant MMP-14 (catalytic domain) (112-298)
Fournisseur: ENZO LIFE SCIENCES
BMLSE2590010
ENZOBMLSE2590010EA 508.2 EUR InStock
ENZOBMLSE2590010
Human recombinant MMP-14 (catalytic domain) (112-298)
Protéines et Peptides
Produced in E.coli. Active Matrix Metalloproteinase-14 (MMP-14, Membrane-Type Matrix Metalloproteinase 1, MT1-MMP) catalytic domain from human cDNA.
The enzyme consists of the catalytic domain of human MMP-14 (Tyr112-Arg298, NM_004995) with a C-terminal purification tag. This represents a naturally-occurring active form of MMP-14 which lacks the C-terminal hemopexin domain.
MMPs lacking this domain cannot cleave native collagens; however, activity toward other targets such as gelatin, casein, or peptide substrates is unaffected.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité