Human recombinant Cathepsin K
Fournisseur: ENZO LIFE SCIENCES
BMLSE5530010
ENZOBMLSE5530010EA 474.1 EUR InStock
ENZOBMLSE5530010
Human recombinant Cathepsin K
Protéines et Peptides
Highly active
Produced in insect cells. Purified as full-length proenzyme (see Prod. No. BML-SE367), then auto-activated at low pH.
Cathepsin K, a member of the papain family of cysteine proteases, acts upon proteins such as collagen, AL amyloid, kinin, and elastin. It is a lysosomal protease expressed primarily in osteoclasts, but also in other cell types such as macrophages. It functions in bone remodeling and is implicated in disease states such as osteoporosis, atherosclerosis, arthritis, and pycnodysostosis.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité