Human recombinant SIRT5 (from E. coli), His-tag
Fournisseur: ENZO LIFE SCIENCES
BMLSE5550050
ENZOBMLSE5550050EA 478.5 EUR InStock
ENZOBMLSE5550050
Human recombinant SIRT5 (from E. coli), His-tag
Protéines et Peptides
SIRT5, along with SIRTs 3 and 4, is one of three human mitochondrial sirtuins (homologs of yeast Sir2). SIRT5, which is localised to both the mitochondrial matrix and the intermembrane space, is an NAD+-dependent lysine deacetylase (class III HDAC).
  • Produced in E. coli
  • Highly active
  • Purified by multi-step chromatography (SDS-PAGE)
  • Useful for studies of enzyme kinetics, modulator activity, drug discovery
It has a liquid formulation in 25 mM TRIS at pH 7,5, 100 mM sodium chloride, 5 mM DTT, containing 10% (v/v) glycerol.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité