Human Recombinant HDAC10 (from Insect cells (Sf9)), HIS Tag
Fournisseur: ENZO LIFE SCIENCES
BMLSE5590050
ENZOBMLSE5590050EA 578.6 EUR InStock
ENZOBMLSE5590050
Human Recombinant HDAC10 (from Insect cells (Sf9)), HIS Tag
Protéines et Peptides
Highly active
Produced in Sf9 cells. HDAC10 from human cDNA, transcript variant 1 (identical to GenBank accession NM_032019) is fused at the C-terminus to a His-tag.
Deacetylates lysine residues on the N-terminal part of the core histones (H2A, H2B, H3 and H4). Histone deacetylation gives a tag for epigenetic repression and plays an important role in transcriptional regulation, cell cycle progression and developmental events.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité