BMLSL1410010
ENZOBMLSL1410010EA
538
EUR
InStock
ENZOBMLSL1410010
Sphingosylphosphorylcholine ≥98% (par HPLC)
Sphingosylphosphorylcholine
Induces release of Ca²⁺ from intracellular stores
Sphingosylphosphorylcholine induces release of Ca²⁺ from intracellular stores via an IP3 independent pathway in permeabilized pancreatic acinar cells. It is an extremely potent mitogen in numerous cell types and induces neurite outgrowth in mouse neuroblastoma cells. It rapidly activates transcription factor AP-14 and transiently activates MAP kinase.
Soluble in water (>50 mg/ml) or ethanol (>50 mg/ml)