Z-Leu-Leu-Leu-vinyl sulfone
Fournisseur: ENZO LIFE SCIENCES
BMLZW91700500
ENZOBMLZW91700500EA 327 EUR InStock
ENZOBMLZW91700500
Z-Leu-Leu-Leu-vinyl sulfone
Protéines et Peptides
Inhibits the trypsin-like, chymotrypsin-like and caspase-like (PGPH) activity in isolated proteasomes and living cells by covalent modification of the amino-terminal threonine of the catalytically active beta subunits. The vinyl sulfone acts as a Michael acceptor for soft nucleophiles such as thiols, leading to the formation of a covalent bond, while being resistant to attack by free thiols, which can cause inactivation of other classes of protease inhibitors.
Inhibits the trypsin-like, chymotrypsin-like and caspase-like (PGPH) activity in isolated proteasomes and living cells by covalent modification of the amino-terminal threonine of the catalytically active beta subunits. The vinyl sulfone acts as a Michael acceptor for soft nucleophiles such as thiols, leading to the formation of a covalent bond, while being resistant to attack by free thiols, which can cause inactivation of other classes of protease inhibitors.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité