NGAL (Dog) ELISA kit
Fournisseur: ENZO LIFE SCIENCES
BPDKIT043
ENZOBPDKIT043EA 1142.9 EUR InStock
ENZOBPDKIT043
NGAL (Dog) ELISA kit
Tests Tests ELISA
A colorimetric immunometric ELISA kit for the rapid measurement of NGAL in dog samples. Absorbance is read at 450 nm.
  • Ready-to-use pre-coated plate and individual calibrators save time and reduce errors
  • Fast results from up to 40 samples in duplicate in just 4 hours per 96 well plate
For the quantitative determination of NGAL in dog urine, plasma, serum, tissue extracts, or culture supernatants.
Information de livraison: Kit consists of microwell plate coated with anti-NGAL (dog) antibody (96 wells), 5X sample diluent, NGAL Dog calibrators 1-8; 0, 4, 10, 20, 40, 100, 200, 400 pg/ml, 25X wash solution concentrate, biotinylated Dog NGAL antibody, HRP-Streptavidin, TMB substrate, stop solution.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité