NGAL (rat) ELISA kit
Fournisseur: ENZO LIFE SCIENCES
BPDKIT046
ENZOBPDKIT046EA 996.6 EUR InStock
ENZOBPDKIT046
NGAL (rat) ELISA kit
Tests Tests ELISA
For the quantitative determination of NGAL in rat urine, plasma, serum, tissue extracts, or culture supernatants.
  • Ready to use pre-coated plate and individual calibrators save time and reduce errors
  • Fast results from up to 40 samples in duplicate in just 4 hours per 96 well plate
  • Absorbance is read at 450 nm
A colorimetric immunometric ELISA kit for the rapid measurement of NGAL in rat samples. Absorbance is read at 450nm.
Information de livraison: The kit contains a microwell plate coated with anti-NGAL (rat) antibody (96 wells), 5X sample diluent, NGAL rat calibrators 1 - 8, 25X wash solution concentrate, biotinylated rat NGAL antibody, HRP-streptavidin, TMB substrate and stop solution.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité