HPV type 16/18/31/33/51 probe, PATHO-GENE®
Fournisseur: ENZO LIFE SCIENCES
ENZ328821000 ENZ328826000
ENZOENZ328821000EA 933 EUR InStock
ENZOENZ328821000 ENZOENZ328826000
HPV type 16/18/31/33/51 probe, PATHO-GENE®
Tests Tests immunologiques
PATHO-GENE® HPV type 16/18/31/33/51 probe is a mixture of biotin-labeled HPV-16, HPV- 18, HPV-31, HPV-33 and HPV-51 specific probes in buffered formamide and hybridisation enhancers.
The PATHO-GENE® HPV type 16/18/31/33/51 probe employs a mixture of the HPV-16, HPV -18, HPV-31, HPV-33 and HPV-51 specific probes that detect HPV types that have been associated with either condyloma or cervical intraepithelial neoplasia (CIN) and carcinoma in situ (CIS). The probe mixture is denatured and hybridised to the tissue section fixed and pretreated on the microscope slide. Specific hybridisation between the HPV DNA probe and DNA in the specimen is determined by the detection of biotin.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité