Green 496 dUTP
Fournisseur: ENZO LIFE SCIENCES
ENZ42831
ENZOENZ42831EA 358.6 EUR InStock
ENZOENZ42831
Green 496 dUTP
Systèmes de liaison et de marquage des protéines
Green 496 [5-Fluorescein] dUTP can replace TTP in reactions in which it serves as a substrate for E. coli DNA polymerase (holoenzyme and Klenow fragment), T4 and Taq DNA polymerases, reverse transcriptase (from AMV and M-MuLV) and terminal transferase.
Fluorescently labeled probes can be prepared with this fluorescent nucleotide by a variety of methods including nick translation, random prime labeling, cDNA labeling and 3’-end labeling. Probes generated by these methods are suitable for use for the identification of specific sequences by in situ hybridisation procedures on fixed cells and tissues by direct fluorescence detection. Green 496 dUTP can also be used for multicolour fluorescence labeling.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité