Gold 525 dUTP
Fournisseur: ENZO LIFE SCIENCES
ENZ42843
ENZOENZ42843EA 472 EUR InStock
ENZOENZ42843
Gold 525 dUTP
Systèmes de liaison et de marquage des protéines
Gold 525 [5(6)-Carboxyrhodamine 6G] dUTP can replace TTP in reactions in which it serves as a substrate for E. coli DNA polymerase (holoenzyme and Klenow fragment), T4 and Taq DNA polymerases, reverse transcriptase (from AMV and M-MuLV) and terminal transferase. Fluorescently labelled probes can be prepared with this fluorescent nucleotide by a variety of methods including nick translation, random prime labelling, cDNA labelling and 3’-end labelling. Probes generated by these methods are suitable for use for the identification of specific sequences by in situ hybridization procedures on fixed cells and tissues by direct fluorescence detection. Gold 525 dUTP can also be used for multicolor fluorescence labelling.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité