DNMT colorimetric drug discovery tests
Fournisseur: ENZO LIFE SCIENCES
ENZ450170001
ENZOENZ450170001EA 1027.4 EUR InStock
ENZOENZ450170001
DNMT colorimetric drug discovery tests
Tests Tests cellulaires
DNMT colorimetric drug discovery kit is a simple method to detect DNA methylation activity or inhibition.
  • Easy colorimetric assay without radioactivity or chromatography
  • Measure as little as 0,5 μg of nuclear extract or 0,5 ng of purified enzymes
  • Optimised for high specificity to 5-mC without cross-reactivity to unmethylated cytosine
  • Reproducible results in less than four hours
Suitable for the quantitative determination of DNMT activity/inhibition in nuclear extracts or purified enzymes.
Information de livraison: Includes wash buffer, DNMT assay buffer, adomet, DNMT enzyme control, capture Ab, detection Ab, enhancer solution, developer solution, stop solution, eight-well assay strips with frame, adhesive covering film.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité