ROS-ID™ ROS/RNS Detection kit
Fournisseur: ENZO LIFE SCIENCES
ENZ51001200
ENZOENZ51001200EA 208 EUR InStock
ENZOENZ51001200
ROS-ID™ ROS/RNS Detection kit
Tests Tests cellulaires
This kit is designed to directly monitor real time reactive oxygen and/or nitrogen species (ROS/RNS) production in live cells using fluorescence microscopy.
Through the combination of the three detection probes with a set of specific inhibitors and activators, this kit enables discrimination among superoxide, nitric oxide and peroxynitrite.
Information de livraison: Kit contains Nitric Oxide Detection Reagent (Red), 60 µL, Oxidative Stress Detection Reagent (Green), 300 nmoles, Superoxide Detection Reagent (Orange), 300 nmoles, Nitric oxide Inducer (L-Arginine), 100 µL, ROS Inducer (Pyocyanin), 1 µmole, Nitric oxide Scavenger (c-PTIO), 400 nmoles, ROS Inhibitor (N-acetyl-L-cysteine), 2 x 10 mg and10X Wash Buffer, 15 mL.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité