ROS-ID™ superoxide detection kit
Fournisseur: ENZO LIFE SCIENCES
ENZ51012
ENZOENZ51012EA 246.4 EUR InStock
ENZOENZ51012
ROS-ID™ superoxide detection kit
Tests Tests cellulaires
This kit is designed to directly monitor real time superoxide production in live cells using fluorescence microscopy and/or flow cytometry.
  • Directly monitors global levels of superoxide in live cells by fluorescence microscopy or flow cytometry
  • High sensitivity, specificity and accuracy for live cell studies
  • Compatible with major components of tissue culture media (phenol red, FBS and BSA)
  • Complete set of reagents, including ROS inducer and scavenger
A major component of the kit, Superoxide Detection Reagent (Orange), is a cell-permeable probe that reacts specifically with superoxide, generating an orange fluorescent product.
The kit is not designed to detect reactive peroxide, hydroxyl, peroxynitrite, chlorine or bromine species, as the fluorescent probe included is relatively insensitive to these analytes. Upon staining, the fluorescent product generated can be visualized using a wide-field fluorescence microscope equipped with a standard orange (e.g., 550/620 nm) fluorescent cube, or cytometrically using any flow cytometer equipped with a blue (488 nm) laser.
The Superoxide Detection Kit contains sufficient reagents for at least 200 microscopy assays or 50 flow cytometry assays using live cells (adherent or in suspension).
Information de livraison: Superoxide Detection Reagent (Orange), 300 nmoles, ROS Inducer (Pyocyanin), 1 µmole, ROS Inhibitor (N-acetyl-L-cysteine), 2 x 10 mg and Wash Buffer Salts, 1 pack.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité