ROS-ID™ NO Detection kit
Fournisseur: ENZO LIFE SCIENCES
ENZ51013200
ENZOENZ51013200EA 246.4 EUR InStock
ENZOENZ51013200
ROS-ID™ NO Detection kit
Tests Tests cellulaires
The kit is designed to directly monitor real time Nitric Oxide production in live cells by fluorescence microscopy using Nitric oxide Detection Reagent (red fluorescent) as the major component.
  • Directly monitor real time NO production in live cells by fluorescence microscopy
  • High specificity, sensitivity and accuracy, yielding a water-insoluble red fluorescent product
  • Distinguish between peroxynitrite and nitric oxide
  • NO inducer and scavenger included
The non-fluorescent, cell-permeable NO detection dye reacts with Nitric Oxide in the presence of O2 with high specificity, sensitivity and accuracy, yielding a water-insoluble red fluorescent product. Importantly, this dye is not reactive towards peroxynitrite that allows distinguishing between peroxynitrite and nitric oxide.
The kit is not designed to detect reactive chlorine or bromine species, as the fluorescent probes included are relatively insensitive to these analytes. Upon staining, the fluorescent products generated by this dye can be visualized using a wide-field fluorescence microscope equipped with any red fluorescent cube. The combination of 650/670 nm is recommended when additional fluorescence signals (green or orange) would be detected simultaneously.
Information de livraison: Nitric Oxide Detection Reagent (Red), 60 µl, Nitric Oxide Inducer (L-Arginine), 100 µl, Nitric Oxide Scavenger (c-PTIO), 400 nmoles and 10X Wash Buffer, 15 ml.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité