LYSO-ID® red cytotoxicity kit (GFP-CERTIFIED®) for microplates
Fournisseur: ENZO LIFE SCIENCES
ENZ51015KP002
ENZOENZ51015KP002EA 471.9 EUR InStock
ENZOENZ51015KP002
LYSO-ID® red cytotoxicity kit (GFP-CERTIFIED®) for microplates
Tests Tests cellulaires
A Rapid, Quantitative and HTS‐compatible Live Cell Cytotoxicity Assay
  • Assay includes unique drug-like dye that rapidly partitions into cells and labels acidic organelles
  • Only commercial assay available that allows for long-term cell monitoring of cytotoxic effects
  • Multi-well, high-throughput with rapid 10-15 minute dye incubation
  • No co-incubation with artificial phospholipid analogs required for detection, eliminating the potential for confounding dye-associated artifacts
  • Monitors lysosome accumulation as a response to prolonged drug treatment
  • Quantitative results in as little as 3 hours, including drug treatment
96-well cell-based assay. Provides a rapid and quantitative approach for determining drug- or toxic agent-induced lysosome and lysosome-like organelle perturbations in live cells.
Informations de commande: Quantity: 2×96-well plates Contents: 10x dual color detection reagent: 2×1 ml
Detection buffer: 20 ml
Verapamil control: 3 µmoles
10x assay buffer: 15 ml
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité