ENZ53001C001
ENZOENZ53001C001EA
83.6
EUR
InStock
ENZOENZ53001C001
Pyocyanin ≥98% (par HPLC)
Pyocyanin
ROS inducer.
Cytotoxic pigment secreted by Pseudomonas aeruginosa, which frequently infects the lungs of immuno-suppressed patients, as well as those with cystic fibrosis. Pyocyanin toxicity results from its ability to undergo reduction by NAD(P)H and subsequent generation of superoxide and H₂O₂ directly in the lungs.
Reconstitute the contents of the vial in 100 µl solvent to yield a 10mM stock solution.
Soluble in DMSO or dimethyl formamide.