Synonyms:
Aflatoxin R0
ENZCHM1040001
ENZOENZCHM1040001EA
725
EUR
InStock
ENZOENZCHM1040001
Aflatoxicol ≥95% (par HPLC)
Aflatoxicol
Mycotoxin. Metabolite of Aflatoxin.
Rare Aflatoxin that is similar but less toxic than Aflatoxin B₁. Exposure may produce necrosis, cirrhosis and carcinoma of the liver. Induces DNA damage. In vitro metabolite of Aflatoxin B₁.
Soluble in DMSO, ethyl acetate, methanol, ethanol, or petrol ether. Not soluble in water.