ENZCHM1660001
ENZOENZCHM1660001EA
140.8
EUR
InStock
ENZOENZCHM1660001
EQ2 NHS ester colorant "quencher" réactif aux amines
EQ2 NHS ester
Amine-reactive quencher dye has a broad visible absorption but no detectable fluorescence emission in fluorescence resonance energy transfer (FRET) applications.
- Commonly used in molecular biology FRET applications to quench fluorophores
- NHS ester quencher dye can be conjugated to amine-labeled probes
- Enable multiplexing of probes with the use of different reporter-quencher dye pairs
- Compatible with most common commercially available fluorophores
EQ2 NHS ester is an amine-reactive quencher dye. It has a broad visible absorption but no detectable fluorescence emission, making it useful as an acceptor in fluorescence resonance energy transfer (FRET) applications.
It can be used to quench reporter dyes: TET, JOE, VIC, HEX, Cy3, NED, TAMRA, Cy3.5, ROX, Texas Red, Cy5, Quasar® 570, Pulsar® 650, CAL Fluor® Gold 540, CAL Fluor® Orange 560, CAL Fluor® Red 590, CAL Fluor® Red 610, CAL Fluor® Red 635, Alexa Fluor® 532, Alexa Fluor® 546, Alexa Fluor® 555, Alexa Fluor® 568, Alexa Fluor® 594, Alexa Fluor® 610, Alexa Fluor® 633, Alexa Fluor® 647, DyLight® 550, DyLight® 594, DyLight® 633, DyLight® 650, ATTO 520, ATTO 532, ATTO Rho6G, ATTO 550, ATTO 565, ATTO Rho11, ATTO 590, ATTO 594, ATTO 633, ATTO Rho14, and ATTO 647.