CYGREEN® Nucleic acid Dye
Fournisseur: ENZO LIFE SCIENCES
ENZGEN1050100
ENZOENZGEN1050100EA 165 EUR InStock
ENZOENZGEN1050100
CYGREEN® Nucleic acid Dye
Colorants Colorants pour gels d'electrophorèse
Nucleic acid dye for gel staining and higher sensitivity qPCR.
  • Higher sensitivity allows detection of low target concentration
  • Greater PCR efficiency for accurate and reproducible results
  • Soluble in water for higher stability
  • Compatible with various enzyme systems
  • Suitable for commonly used rotor-type and plate-type qPCR platforms
  • Validated for qPCR applications and gel staining
  • Lower cost per assay compared to commonly used dyes
CYGREEN® Nucleic acid dye is a DNA intercalating agent that is used to stain DNA. The DNA-dye complex emits a fluorescence spectra that makes it suitable for qPCR and gel staining applications. CYGREEN® Nucleic acid dye has excitation/emission spectra similar to commonly used qPCR dyes and can be substituted in to increase sensitivity and reduce cost.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité