alphaRsaalphaI
Fournisseur: ENZO LIFE SCIENCES
ENZGEN1091000
ENZOENZGEN1091000EA 83.6 EUR InStock
ENZOENZGEN1091000
alphaRsaalphaI
Protéines et Peptides
Restriction  enzyme for molecular biology applications
  • High quality enzyme with stringent QC testing
  • Includes 10X Cutting Buffer
  • BSA premixed into buffers
  • Suitable for molecular cloning, restriction site mapping, genotyping, southern blotting, SNP
Produced in an E. coli strain that carries the cloned RsaI gene from Rhodopseudomonas sphaeroides.
RsaI is a restriction enzyme that recognizes GT^AC sites.10X Cutting Buffer is included that contains BSA, which enhances enzyme stability and binds to contaminants in DNA preps.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité