Absorbance 96 Plate Reader
Fournisseur: ENZO LIFE SCIENCES
ENZINSA96
ENZOENZINSA96EA 5240 EUR InStock
ENZOENZINSA96
Absorbance 96 Plate Reader
Lecteurs de microplaques
The Absorbance 96 is a uniquely designed microplate reader for assays in 96-well format. By far the most compact reader on the market, the footprint of the plate reader is almost as small as the plate itself.
  • Space saving design
  • Fast readout speed
  • Reliable measurement results
  • Maintenance-free
  • Solid state technology
The Absorbance 96 delivers precise, accurate results at an affordable price. Together with its proprietary software, the Absorbance 96 is ideal for a variety of applications extending from ELISA to protein- and cell-based assays.
Connectivity/communication: Through USB connection 5 V
Detection method: Absorbance
Shipping: Ambient temperature
Attention: Research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité