Adipogenesis assay kit
Fournisseur: ENZO LIFE SCIENCES
ENZKIT1030005
ENZOENZKIT1030005EA 421.3 EUR InStock
ENZOENZKIT1030005
Adipogenesis assay kit
Tests Tests cellulaires
Convenient method for the enhanced induction and detection of adipogenesis, in the classic 3T3L-1 model.
  • Provides optimized reagents for adipocyte differentiation and subsequent colorimetric lipid oil detection
  • Contains enhancer solution for the optimal induction of adipogenesis
  • For drug-screening and testing agonists/antagonists of adipogenesis
For the induction and colorimetric detection of adipogenesis.
The Adipogenesis Assay Kit provides all the reagents needed to induce and detect adipogenesis. Optimized Differentiation, Insulin, and Enhancer solutions ensure maximal adipocyte induction. The Adipogenesis Dye allows for visual confirmation of differentiation and can also be easily extracted allowing measurement using a spectrophotometer or microplate reader.
Contains enough reagents to conduct experiments for: 5 x 24 well plates or 225 cm² tissue culture plate surface area.
Informations de commande: Contains 1000x Differentiation Solution (Dexamethasone and IBMX), 1000x Insulin Solution, 1000x Enhancer Solution (Rosiglitazone), 1x Cell Fixative, 1x Adipogenesis Dye (Oil Red O), 1x Extraction Solution.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité