LH ELISA kit, grow cells
Fournisseur: ENZO LIFE SCIENCES
ENZKIT1070001
ENZOENZKIT1070001EA 316.8 EUR InStock
ENZOENZKIT1070001
LH ELISA kit, grow cells
Tests Tests ELISA
Highly sensitive LH ELISA kit enabling detection of LH in serum, plasma and tissue culture media in just 2 hours.
  • Sensitive measurement of LH, detecting as little as 5,2 mIU/ml
  • Negligible reactivity from similar glycoprotein hormones
  • High throughput format with results in two hours for up to 39 samples in duplicate
  • Fully quantitative results that surpass semi-quantitative Western blot analysis
For the quantitative determination of LH in tissue culture media, serum and plasma in human and rat samples.
The LH ELISA kit is a colorimetric competitive enzyme immunoassay kit with results in two hours.
Informations de commande: Contains Microtiter Plate, Assay Buffer 13, Standard, Wash Buffer Concentrate, Antibody, Tracer, Conjugate, Substrate, and Stop Solution.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité