FSH ELISA kit, Grow Cells
Fournisseur: ENZO LIFE SCIENCES
ENZKIT1080001
ENZOENZKIT1080001EA 295.9 EUR InStock
ENZOENZKIT1080001
FSH ELISA kit, Grow Cells
Tests Tests ELISA
Highly sensitive FSH ELISA kit enabling detection of FSH in serum, plasma and tissue culture media in just 2 hours.
  • Sensitive measurement of FSH, detecting as little as 0,5 mIU/ml
  • Negligible reactivity from similar glycoprotein hormones
  • High throughput format with results in two hours for up to 37 samples in duplicate
  • Fully quantitative results that surpass semi-quantitative Western blot analysis
For the quantitative determination of FSH in tissue culture media, serum and plasma in human samples.
The FSH ELISA kit is a colorimetric sandwich immunoassay kit with results in two hours.
Informations de commande: Contains Microtiter Plate, Assay Buffer 13, Standard, Wash Buffer Concentrate, Antibody, Tracer, Conjugate, Substrate, and Stop Solution.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité