Cellular senescence activity assay
Fournisseur: ENZO LIFE SCIENCES
ENZKIT1290120
ENZOENZKIT1290120EA 673.2 EUR InStock
ENZOENZKIT1290120
Cellular senescence activity assay
Tests Tests cellulaires
Highly reproducible SA-β-gal activity kit
  • Quantify SA-β-gal activity in a fluorescence plate reader
  • Easy-to-use protocol with results in less than 90 minutes
  • Fully compatible with all cell types
For the determination of cellular senescence by measuring SA-β-gal activity using a fluorometric substrate. The 96-well Cellular senescence assay kit provides an easy-to-use and efficient method to determine the cellular senescence by measuring SA-β-gal activity using a fluorometric substrate. This quantitative assay uses cell lysate for both SA-β-galactosidase activity determination and normalization of samples containing different cell numbers. Each kit provides sufficient quantities to perform up to 120 assays in a 96-well plate.
Informations de commande: Contains 2X Cell lysis buffer, 2X Reaction buffer, SA-?-gal substrate (20X), Stop solution.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité