Methotrexate ELISA kit
Fournisseur: ENZO LIFE SCIENCES
ENZKIT1420001
ENZOENZKIT1420001EA 735 EUR InStock
ENZOENZKIT1420001
Methotrexate ELISA kit
Tests Tests ELISA
Ultra sensitive Methotrexate ELISA kit enabling detection of Methotrexate in serum, plasma, and urine in just 1,5 hours.
  • Sensitive measurement of methotrexate, detecting as little as 0,087 ng/ml
  • Broad dynamic range suitable for a large variety of samples
  • High throughput format with results in 1,5 hours for up to 38 samples in duplicate
  • Fully quantitative results that surpass semi-quantitative Western blot analysis
For the quantitative determination of methotrexate in serum, plasma and urine in human, mouse, and rat samples. The Methotrexate ELISA kit is a colorimetric competitive immunoassay kit with results in 1,5 hours.
Informations de commande: Contains Microtiter Plate, Assay Buffer 13, Standard, Wash Buffer Concentrate, Antibody, Antibody Diluent, Conjugate, Substrate, and Stop Solution.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité