Human growth hormone ELISA kit
Fournisseur: ENZO LIFE SCIENCES
ENZKIT1480001
ENZOENZKIT1480001EA 630 EUR InStock
ENZOENZKIT1480001
Human growth hormone ELISA kit
Tests Tests ELISA
Ultra-sensitive human growth hormone ELISA kit enabling detection of HGH in serum, plasma, saliva and tissue culture media in just 2,5 hours.
  • Ultra-sensitive measurement of HGH, detecting as little as 0,93 pg/ml
  • Negligible cross reactivity with similar proteins (prolactin, human placental lactogen)
  • High throughput format with results in 2,5 hours for up to 36 samples in duplicate
  • Fully quantitative results that surpass semi-quantitative Western blot analysis
For the quantitative determination of growth hormone in serum, plasma, saliva and tissue culture media. The human growth hormone ELISA kit is a colorimetric immunometric immunoassay kit with results in 2,5 hours.
Informations de commande: Contains Microtiter plate, Assay Buffer 13, Standard, Wash Buffer Concentrate, Antibody, Antibody Diluent, Conjugate, Substrate,  and Stop Solution.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité