Research® Plus G Basic Single-Channel, Variable, Volume Pipette
Fournisseur: EPPENDORF
3123000.349
EPPE3123000.349EA 245 EUR InStock
EPPE3123000.349
Research® Plus G Basic Single-Channel, Variable, Volume Pipette
Pipettes Micropipettes
The Eppendorf Research plus is one of the safest and most ergonomic mechanical pipettes currently available. It not only meets the highest requirements for precision and accuracy, but also offers a high degree of flexibility thanks to a large selection of variants. This makes the Research plus an indispensable companion in your laboratory. Thanks to its remarkably low weight and reduced effort when pipetting, this micropipette helps to relieve the strain on your hand and arm.
  • Adjustable
  • Manual
  • Single-channel variable
  • Without tip box
Depending on your needs and application, autoclave the entire pipette or just the base to ensure proper decontamination. High-quality material ensures high UV and chemical resistance; special single-channel bases available for aggressive chemicals. Single-channel pipettes with ACT label and epT.IPS pipette tips help you make an informed choice when it comes to sustainability.
Features and benefits:
Mechanical air cushion pipette with three-button operation as standard (one control for volume selection, one for pipetting and one for tip ejection)
Ideal for accurate, easy and ergonomic pipetting of aqueous solutions by hand
Convince yourself of the difference in weight and effort required when pipetting: Developed according to the principles of the Eppendorf PhysioCare Concept
Benefit from one of the pipettes with the lowest operating and tip ejection forces on the pipette market to reduce hand and arm strain
Minimised attachment forces thanks to a spring-loaded tip cone (available for all pipettes up to 1 ml)
Adjust your pipette in seconds to improve accuracy when pipetting difficult liquids such as ethanol or when pipetting at high altitudes. Then return to factory adjustment without calibration
Application:
Forward pipetting
Reverse pipetting
Removal of excess material
Sample mixing
Phase extraction
Filling of plates, gels and reaction vessels
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité