Thermocycleurs, Mastercycler® ep
Fournisseur: EPPENDORF
5345000.013 5341000.043 5341000.540 5344000.044 5344000.540 5349810.001
732-0124EA 0 EUR InStock
732-0124 732-0909 732-0411 732-0915 732-0413 EPPE5349810.001
Thermocycleurs, Mastercycler® ep
Thermocycleurs PCR
The Mastercycler® ep family of thermal cyclers are distinguished by having thermoblocks with extremely fast heat-ing and cooling speeds, and unique heated lid technology (ESP - Electronic Sample Protection), which avoids the preheating of samples and minimises unspecific annealing. The gradient option of each Mastercycler® ep system features SteadySlope® technology to ensure that the heating and cooling ramp rates are identical in both gradient and normal operation. Programming with the control panel is easy and intuitive, allowing novice users to get started quickly and safely.
  • Control panel or PC software driven
  • ESP heated lid
  • Available with manual or motorised lid
  • Space-saving footprint with sliding lid and no side air vents necessary
  • SteadySlope® technology ensures identical temperature control characteristics in both optimisation and rou-tine experiments, as well as a reliable transfer of the optimisation results to the routine application
For integration into fully automated environments, Mastercycler® ep models are also available with an optional motorised lid. For additional speed, the Mastercycler® ep gradient S incorporates a device driven Hot Start. The Mastercycler® ep models can be operated in various configurations and adjusted to the requirements and size of the laboratory - as a universal, stand alone device with control panel operation; as a ‘mini-satellite’ system consisting of a control panel and up to 5 Mastercycler ep models; as a PC supported, GLP conforming ‘PCR machine’; as a PC controlled Mastercycler ep network; or as high throughput units with motorised lids for automation integration.
Temperature control range of block: 4 to 99 °C
Interfaces: Centronics®, RS232, control panel, CAN_in/CAN_out
* Un panneau de commande ou un logiciel epCycleManager est nécessaire au fonctionnement; des câbles de connexion CAN_Bus sont requis pour connecter les cycleurs en réseau.
Informations sur les accessoires: L'adaptateur in situ permet de convertir le Mastercycler® ou Mastercycler® gradient en appareil in situ. Il est fourni avec un capot transparent.
Commander maintenant
COMMANDE
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité