Heat sealers, Eppendorf HeatSealer S100/S200
Fournisseur: EPPENDORF
5391000.001 5391000.036 5392000.005 5392000.030
EPPE5391000.001EA 0 EUR InStock
EPPE5391000.001 EPPE5391000.036 EPPE5392000.005 EPPE5392000.030
Heat sealers, Eppendorf HeatSealer S100/S200
Thermosoudeuses Thermosoudeuses pour microplaques
The Eppendorf HeatSealer Family offers reliable and effective sealing of a variety of plates. When working with plates, only a tight and reproducible seal gives predictable and reproducible assay performance. Every laboratory works with different kinds of plates and the HeatSealer will work with most common formats — from large deepwell plates to low profile PCR plates. Specific adapter sets are available to cover almost any plate format.
  • Hermetic sealing of multiwell plates
  • Minimises evaporation in PCR, reducing cross contamination
  • Seals 96- and 384-well
More Product Information
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité