VOA vials, economy certified
Fournisseur: EP SCIENTIFIC
GVB-100C
EPSCGVB-100CEA 195 EUR InStock
EPSCGVB-100C
VOA vials, economy certified
Fioles
Borosilicate glass, clear. Septa vials with all the essentials and none of the frills. The VOA vials are processed and certified to meet EPA Performance based specifications for volatile organic analysis. Boxed with a protective chip board divider to prevent breakage during transport. Vial shoulder slope optimized to prevent air bubble entrapment. Comes with open-top white polypropylene cap PTFE-lined silicone septum which is permanently bonded to the cap for a secure seal. Certificate of analysis included.
Processed to meet or exceed EPA performance based specifications for volatile organic analysis.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité