Bottles, narrow neck
Fournisseur: EP SCIENTIFIC
S349-1000
EPSCS349-1000EA 124 EUR InStock
EPSCS349-1000
Bottles, narrow neck
Flacons Flacons en verre
Type III soda-lime glass bottle.
  • Certified bottles meet or exceed EPA performance based standards for volatile organic analysis
  • Manufactured under an ISO certified quality management system
  • Amber glass protects light-sensitive chemicals
Narrow-neck glass bottle with septa closure is designed for VOA testing. Equipped with a pierceable PTFE-faced silicone septum. Cap features a moulded-in septa retainer ring which holds the septa securely in place without bonding to the cap so septum can be removed and replaced as needed (except 1 L bottle).
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité