Cleanroom wipes, C3
Fournisseur: ESSENTRA POROUS TECHNOLOGIES
7-C3-99L-00
EPTA7-C3-99L-00EA 1130 EUR InStock
EPTA7-C3-99L-00
Cleanroom wipes, C3
Lingettes Lingettes pour salles propres Lingettes pour salles propres, sèches
These 100% hydroentangled polyester wipes offer the highest level of softness and abrasion resistance − perfect for cleaning and polishing the most sensitive surfaces.
  • Tough and absorbent for usage in a variety of applications
  • Recommended for the cleaning of lenses or other sensitive surfaces
  • Chemically compatible with most cleaning solutions
  • Low NVRs and ions
  • No chemical binders
Typical applications include: Optics cleaning and manufacturing, LCD and display cleaning and maintenance, sensitive surface wipe downs and prep, use as a work surface for delicate tasks, critical tool and component wipe downs.
Recommended for use in ISO 5 - 8 controlled environments.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité