ridoMED Magnifying Spectacles
Fournisseur: ESCHENBACH OPTIK
16363
ESCH16363EA 402 EUR InStock
ESCH16363
ridoMED Magnifying Spectacles
Loupes
Magnifying spectacles for near viewing.
  • Coated lens system
  • Achromatic lens made of high quality silicate
  • Adjustment of pupillary distance of 54 to 74 mm
  • Lens diameter 23 mm
  • Visual field near: 55 mm
Focusing/dioptre compensation adjusted individually for each eyepiece. Ocular-side additional correction is possible, e.g. cylinder correction. Sturdy carrier frame made from glass fibre reinforced, black plastic. Comfortable, easily adjustable saddle bridge. Supplied in black zippered case. With convergence angles to suit the respective working distances.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité