Temperature and humidity data logger, iLog
Fournisseur: Askey Dataloggers
ED-EI-HS-D-32L
ESCOED-EI-HS-D-32LEA 438 EUR InStock
ESCOED-EI-HS-D-32L
Temperature and humidity data logger, iLog
Enregistreurs de données
The iLog data loggers are designed for the monitoring of both temperature and humidity on sensitive products while supporting compliance requirements. It can be start with magnetic key swipe or at pre-defined time and stopped after pre-defined readings or timer.
  • Large LCD display with multi-functional viewing
  • Visual and audible alarms
  • Bookmark capabilities
  • Wide temperature range (high heat, ambient, refrigerated, frozen and dry ice)
  • Password protected
Certifications: IP 65 rated protection class.
Information de livraison: Product comes with two years of warranty.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité