Binocular stereo microscope StereoBlue 2x/4x revolving objective, 20x/40x magnification with ergonomical pillar stand with incident and transmitted LED illumination 1 * 1 Pce
Fournisseur: EUROMEX
Binocular stereo microscope StereoBlue 2x/4x revolving objective, 20x/40x magnification with ergonomical pillar stand with incident and transmitted LED illumination 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Binocular stereo microscope StereoBlue 2x/4x revolving objective, 20x/40x magnification with ergonomical pillar stand with incident and transmitted LED illumination 1 * 1 Pce EUMXSB.1402-P 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou