Trinocular stereo zoom microscope StereoBlue, 0.7x to 4.5x zoom objective, magnification from 7x to 45x, ergonomical pillar stand with incident and transmitted LED illumination A pair of WF10x/20mm wide field eyepieces with eyecups Trinocular head wi 1 * 1 Pce
Fournisseur: EUROMEX
Trinocular stereo zoom microscope StereoBlue, 0.7x to 4.5x zoom objective, magnification from 7x to 45x, ergonomical pillar stand with incident and transmitted LED illumination A pair of WF10x/20mm wide field eyepieces with eyecups Trinocular head wi 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Trinocular stereo zoom microscope StereoBlue, 0.7x to 4.5x zoom objective, magnification from 7x to 45x, ergonomical pillar stand with incident and transmitted LED illumination A pair of WF10x/20mm wide field eyepieces with eyecups Trinocular head wi 1 * 1 Pce EUMXSB.1903-P 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou