Antimicrobial extra foam hand cleanser, OxyBAC®
Fournisseur: Deb-STOKO
OXYEX1LBG
EVONOXYEX1LBGEA 152 EUR InStock
EVONOXYEX1LBG
Antimicrobial extra foam hand cleanser, OxyBAC®
Soin de la peau Savons et soins nettoyants
Antimicrobial rich-cream foam hand wash with OptiDose™ pump, perfume-free and dye-free.
  • Contains accelerated hydrogen peroxide
  • Skin hypoallergenic
  • Suitable for food facilities and healthcare workers
  • Excellent skin cleansing
  • Hygienic cartridge
Dispenses foam on the hands which is instantly ready for washing and quick & easy rinsing from hands and sinks. Helps to prevent drying and leave the skin feeling smooth after use.
Certifications: HACCP Certified. Passes EN1499 just with ONE dose.
Attention: Use biocides safely. Always read the label and product information before use.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité