Hand disinfectants, Swarfega® Orange
Fournisseur: Deb-STOKO
SOR2LT SORC4LTR
EVONSOR2LTEA 114 EUR InStock
EVONSOR2LT EVONSORC4LTR
Hand disinfectants, Swarfega® Orange
Soin de la peau Désinfectants Mousses et liquides désinfectants
This is a solvent-free workshop cream with plastic microbeads, effective against strong and adherent soiling such as greases, oils and fuels.
  • Glycerin and wheat germ oil increase skin tolerance and hydrate the skin
  • Fresh fragrance based on natural orange extracts
  • Solvent-free form helps to prevent skin dryness
  • Contains glycerin and wheat germ extracts to help hydration of cutaneous skin
Contains micropolymer granules which provide deep cleaning and gently loosen crusted contaminants without damaging the skin. This rich and creamy lotion rinses offMore Product Information
Collapse All
Désignation Cdt Numéro de catalogue VWR Quantité
DEB Swarfega® Orange Wash, cartridge, 2 L 6 EVONSOR2LT
DEB Swarfega® Orange Wash, cartridge, 4 L 4 EVONSORC4LTR
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité