Avantor® ACE® Excel® C8, HPLC/UHPLC Columns, Analytical, 5 µm
Fournisseur: Avantor
EXL-122-1046UEA 707 EUR InStock
EXL-122-1046U EXL-122-1246U EXL-122-1546U EXL-122-0202U EXL-122-0246U EXL-122-2502U EXL-122-2546U EXL-122-0502U EXL-122-0503U
Avantor® ACE® Excel® C8, HPLC/UHPLC Columns, Analytical, 5 µm
Colonnes pour chromatographie
These ultra-inert Avantor® ACE® Excel® C8 columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (2 to 5 µm) and complementary phases
  • Can be used as general purpose starting point column
  • Method validation kits contain three different batches of the same phase - available on request
These columns are designed for a wide range of chromatographic applications, to provide great performance with acidic, basic and neutral molecules.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité