Avantor® ACE® Excel® C18, HPLC/UHPLC Columns, Analytical, 1,7 µm
Fournisseur: Avantor
EXL-171-7503UEA 953 EUR InStock
EXL-171-7503U EXL-171-1003U EXL-171-1002U EXL-171-0503U EXL-171-0502U EXL-171-0303U EXL-171-0302U EXL-171-0202U EXL-171-D1002
Avantor® ACE® Excel® C18, HPLC/UHPLC Columns, Analytical, 1,7 µm
Colonnes pour chromatographie
Avantor® ACE® C18 columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (1.7 to 5 µm) and complementary phases
  • Method validation kits contain three different batches of the same phase - available on request
These columns are designed for a wide range of chromatographic applications, to provide great performance with acidic, basic and neutral molecules.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité