Avantor® ACE® Excel® C18-Amide, HPLC/UHPLC Columns, Analytical, 1,7 µm
Fournisseur: Avantor
EXL-1712-7502UEA 952 EUR InStock
EXL-1712-7502U EXL-1712-1003U EXL-1712-1002U EXL-1712-0503U EXL-1712-0502U EXL-1712-0302U EXL-1712-0202U ACE-1P12-GD2U EXL-1712-D1002
Avantor® ACE® Excel® C18-Amide, HPLC/UHPLC Columns, Analytical, 1,7 µm
Colonnes pour chromatographie
Avantor® ACE® Excel® C18-Amide is a uniquely designed stationary phase offering improved retention of polar compounds, enhanced phenolic resolution and alternative selectivity to C18 bonded columns. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (1,7 to 5 µm) and complementary phases
  • Low bleed for UV and LC/MS compatibility
  • C18 hydrophobic plus Amide interactions, compatible with 100% aqueous condition
  • Method validation kits contain three different batches of the same phase - available on request
The special design of the Avantor® ACE® C18-Amide ligand incorporates an extended alkyl chain spacer between the silica surface and amide polar group, which gives extra phase stability.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité