Avantor® ACE® Excel® CN-ES, HPLC/UHPLC Columns, Analytical, 1,7 µm
Fournisseur: Avantor
EXL-1713-7503UEA 0 EUR InStock
EXL-1713-7503U EXL-1713-7502U EXL-1713-1003U EXL-1713-1002U EXL-1713-0503U EXL-1713-0502U EXL-1713-0303U EXL-1713-0302U ACE-1P13-GD2U EXL-1713-D1002
Avantor® ACE® Excel® CN-ES, HPLC/UHPLC Columns, Analytical, 1,7 µm
Colonnes pour chromatographie
Avantor® ACE® CN-ES combines CN polar selectivity with enhanced hydrophobicity, enabling the benefits of both interactions to be fully exploited. Extra phase stability and increased column lifetime is provided by an extended alkyl chain spacer between the silica surface and cyano group. Avantor® ACE® CN-ES is suitable for both reversed phase and normal phase separations.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • With hydrophobic plus CN polar selectivity, especially useful for polar molecules
  • Wide range of particle sizes (1.7 µm to 10 µm) and complementary phases
  • Method validation kits contain three different batches of the same phase - available on request
These stainless steel columns are available in a wide range of dimensions from capillary to preparative for HPLC and UHPLC separations.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité