Avantor® ACE® Excel® C18-HL, HPLC/UHPLC Columns, Analytical, 5 µm
Fournisseur: Avantor
EXL-321-2546UEA 937 EUR InStock
EXL-321-2546U
Avantor® ACE® Excel® C18-HL, HPLC/UHPLC Columns, Analytical, 5 µm
Colonnes pour chromatographie
Avantor® ACE® Excel® C18-HL columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (3 and 5 µm) and complementary phases
  • Especially suited for preparative work due to higher carbon load
  • Method validation kits contain three different batches of the same phase -
More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité