BIOFLOAT™ 6-well Cell Culture Spheroid Plate (F-bottom)
Fournisseur: faCellitate
BIOFLOAT™
738-0260EA 186 EUR InStock
738-0260
BIOFLOAT™ 6-well Cell Culture Spheroid Plate (F-bottom)
Microplaques Microplaques pour culture cellulaire
Ready-to-use for suspension and 3D cell culture. The BIOFLOAT™ 6-well plate provides a highly defined, fully inert, and cell-repelling surface.
  • Stability in long term cultivation of 3D cell models
  • Scratch resistant
  • Repellent to protein and hydrophobic compounds
  • Cell repellent
  • Optimized for organoids
The BIOFLOAT™ 6-well f-bottom plate provides a highly defined, fully inert, and cell-repelling surface.
Temperature range: – 20 to + 50 °C
Certifications: The product does not use any raw materials of animal or biological origin and therefore does not have TSE/BSE.
The polymer raw materials used for coating plates were positively evaluated for quality consistency using GPC, GC-MS and NMR spectroscopy.
The sterile, non-pyrogenic/endotoxin-free, non-cytotoxic, DNase-/RNase-/DNA-free multi-well plates were coated using pipetting robots. Coated plates were sterilised by electron beam irradiation.
Conditionnement: Bag of 1 plate per pack.
Attention: Research use only. Not intended for diagnostic or therapeutic use.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité