BIOFLOAT™ 384-well Cell Culture Spheroid Plate (U-bottom)
Fournisseur: FACELLITATE
F224384-6
FCELF224384-6EA 1390 EUR InStock
FCELF224384-6
BIOFLOAT™ 384-well Cell Culture Spheroid Plate (U-bottom)
Microplaques Microplaques pour culture cellulaire
The BIOFLOAT™ 96-well plate outperforms existing products by its anti-adhesive properties.
  • Rapid: Spheroids are formed very fast
  • Round: The spheroids are highly uniform and allow for perfect data consistency
  • Reliable: The reliable quality of the BIOFLOAT™ surface makes it a premium product for spheroid and organoid culture
The BIOFLOAT™ 96-well plate provides a highly defined, fully inert, and cell-repelling surface.
Temperature range: – 20 to + 50 °C
Certifications: The product does not use any rawmaterials of animal or biological origin andMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité