Sliding microtome, Slide 2003
Fournisseur: PFM Medical GmbH
030002
FEAT030002EA 0 EUR InStock
FEAT030002
Sliding microtome, Slide 2003
Microtomes et groupes de microtomes Microtomes
The stable knife sliding action has unrivalled smoothness due to the horizontal cross-roller bearing. This results in less fatigue during daily use. The vertical object movement uses a cross-roller bearing system, is free from backlash and avoids vibration. The newly developed quick release system for both the universal cassette clamp and the standard object clamp in connection with the object orientation system and the user-friendly rotatable knife holder allows a quick and uncomplicated approach of the object to the blade. The section thickness can be set manually or automatically. TheMore Product Information
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité