Transfection reagent, TurboFect in-vivo
Fournisseur: Thermo Fisher Scientific
R0541
FERMR0541EA 514 EUR InStock
FERMR0541
Transfection reagent, TurboFect in-vivo
Réactifs de transfection
TurboFect in vivo Transfection Reagent is highly effective for DNA plasmid delivery in vivo, while not inducing an inflammatory response. TurboFect in vivo has been assessed for plasmid DNA delivery in both mice and rats and is suitable for DNA administration via various routes, including intravenous, intraperitoneal, and intratumoral injection. The amount of DNA and suggested injection volume will depend on the experimental animal and the route of administration as well as the targeted tissue or organ and the expression vector.
  • No detectable inflammatory response
  • Successful plasmid delivery via multiple routes of administration
  • Functionally tested in mice and rats
TurboFect in vivo is a sterile solution of a cationic polymer in water. The polymer forms compact, stable, positively charged complexes with DNA. These complexes protect DNA from degradation and facilitate gene delivery in vivo.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité